Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci Human Heart Lysate
SDP

Catalog No. 16916783 Shop All ProSci Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-916-783 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-916-783 Supplier ProSci Supplier No. 13010100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Total protein normal heart - Human

  • Heart tissue lysate was prepared by homogenization in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7.4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0.1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin).
  • Tissue and cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay.
  • The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 50 mM DTT.

Specifications

Quantity 0.1 mg
Storage Buffer Heart lysate is supplied in SDS sample buffer containing 50 mM DTT.
Tissue Condition Normal
Host Species Human
Concentration 2 mg/mL
Product Type Lysate
Content And Storage Store at 2°C to 8°C for continuous use. For extended storage, freeze working aliquots at -70°C. Repeated freezing and thawing is not recommended. Under proper storage conditions the shelf life is half a year from the date of receipt.
Protein State Denatured
For Use With (Application) Western Blot
Tissue Heart
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.