Learn More
Creative Biomart AG-702, HSPs
Supplier:  Creative Biomart THP0198
Encompass_Preferred
The product is a recombinant human heat shock protein also known as stress protein. It is also known as HSPs, which are a group of proteins that are induced when a cell undergoes various types of environmental stresses like heat, cold and oxygen deprivation. HSPs are present in all cells in all life forms from bacteria to mammals, and their structure and function are similar across these diverse life forms.
Catalog No. 50-208-4249
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.