Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Hematopoietic Lineage Antibody Cocktail, APC, eBioscience™
GREENER_CHOICE

Catalog No. 5015419
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-154-19 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-154-19 Supplier Invitrogen™ Supplier No. 22777672
  • $846.00
    $634.65 / Each of 1

    Save $211.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Antibody Cocktail

Description: The Human Hematopoietic Lineage APC Cocktail contains APC-conjugated antibodies that can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on their expression of cell surface antigens. This cocktail contains the following antibodies: Anti-Human CD2 (RPA-2.10) APC Anti-Human CD3 (OKT3) APC Anti-Human CD14 (61D3) APC Anti-Human CD16 (CB16) APC Anti-Human CD19 (HIB19) APC Anti-Human CD56 (TULY56) APC Anti-Human CD235a (HIR2) APC Applications Reported: The Human Hematopoietic Lineage APC Cocktail has been reported for use in flow cytometric analysis. Applications Tested: The Human Hematopoietic Lineage APC Cocktail has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 20 μL per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

Hematopoietic Lineage antibodies can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on their expression of cell surface antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Hematopoietic Lineage
Applications Flow Cytometry
Classification Cocktail
Clone RPA-2.10, OKT3, 61D3, CB16, HIB19, TULY56, HIR2, OKT3, 61D3, CB16, HIB19, TULY56, HIR2
Concentration 20 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias 17A2; Hematopoietic Lineage Cocktail; M1/70; RA3-6B2; RB6-8C5; TER-119
Host Species Mouse
Purification Method Affinity chromatography
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.