Learn More
Invitrogen™ Human Hematopoietic Lineage Antibody Cocktail, FITC, eBioscience™

Description
Description: The human hematopoietic lineage flow cocktail contains FITC-conjugated antibodies that can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on their expression of cell surface antigens. This cocktail contains the following antibodies: Anti-Human CD2 (RPA-2.10) FITC Anti-Human CD3 (OKT3) FITC Anti-Human CD14 (61D3) FITC Anti-Human CD16 (CB16) FITC Anti-Human CD19 (HIB19) FITC Anti-Human CD56 (TULY56) FITC Anti-Human CD235a (HIR2) FITC Applications Reported: Human Hematopoietic Lineage FITC Cocktail has been reported for use in flow cytometric analysis. Applications Tested: Human Hematopoietic Lineage FITC Cocktail has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 20 μL per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Specifications
Specifications
| Antigen | Human Hematopoietic Lineage |
| Applications | Flow Cytometry |
| Classification | Cocktail |
| Clone | RPA-2.10, OKT3, 61D3, CB16, HIB19, TULY56, HIR2, OKT3, 61D3, HIB19, TULY56, HIR2 |
| Concentration | 20 μL/Test |
| Conjugate | FITC |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene Alias | 17A2; Hematopoietic Lineage Cocktail; M1/70; RA3-6B2; RB6-8C5; TER-119 |
| Host Species | Mouse |
| Purification Method | Affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.