Learn More
Immunology Consultants Human Hemopexin ELISA Kit
SDP
Supplier:  Immunology Consultants E80HX
Encompass_Preferred
The Human Hemopexin (HX) test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring HX in human biological samples.
Catalog No. 50-240-6600
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.