Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5057
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5057 Supplier R&D Systems Supplier No. AF5057
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 Polyclonal specifically detects Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Heparan Sulfate 6-O-Sulfotransferase 1/HS6ST1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. AAY14736
Gene Alias DKFZp547H098, EC 2.8.2, EC 2.8.2.-, FLJ25392, heparan sulfate 6-O-sulfotransferase 1, heparan-sulfate 6-sulfotransferase, HS6ST-1, HS6STheparan-sulfate 6-O-sulfotransferase 1, MGC116899, MGC116901
Gene Symbols HS6ST1
Host Species Sheep
Immunogen CHO cell-derived recombinant human HS6ST1 Pro28-Trp401 Accession # AAY14736
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9394
Test Specificity Detects human HS6ST1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse HS6ST3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.