Learn More
American Research Products Inc Human Heparanase 2 HPSE2 ELI
Supplier:  American Research Products Inc CSBEL010717HU
Encompass
Human Heparanase-2 (HPSE2) ELISA kit from CUSABIO Detection Range 0 312 U/mL-20 U/mL Sensitivity 0 078 U/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26971
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.