Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Hepcidin Quantikine ELISA Kit

Catalog No. DHP250
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DHP250 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DHP250 Supplier R&D Systems™ Supplier No. DHP250
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Hepcidin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Hepcidin Quantikine ELISA Kit quantifies human Hepcidin Antimicrobial Peptide in cell culture supernates (25 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (10 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 3.81pg/mL
Conjugate HRP
Cross Reactivity <0.5% cross-reactivity observed with available related molecules. <50% cross-species reactivity observed with species tested
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural Human Hepcidin
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (25uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym HAMP, HEPC, hepcidin antimicrobial peptide, HEPCPutative liver tumor regressor, HFE2B, HFE2Bhepcidin, LEAP1, LEAP-1, LEAP1PLTR, Liver-expressed antimicrobial peptide 1, PLTR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.