Learn More
ATCC Human herpesvirus 5; Strain: UL32-EGFP-HCMV-TB40
Supplier:  ATCC VR3348
Encompass
Human herpesvirus 5 strain UL32-EGFP-HCMV-TB40 is propagated in MRC-5 cells (ATCC CCL-171). This strain was sourced from the in vitro recombination of HCMV strain TB40 with a plasmid carrying the TB40 UL32 gene fused to GFP. Because GFP is associated with a structural protein, virus particles fluoresce green under appropriate illumination; this is useful for following intracellular traffic of virus particles.
Catalog No. NC2235214
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.