Learn More
ATCC Human herpesvirus 5 DNA; HCMV strain Towne
Supplier:  ATCC VR977D
Encompass
Viral nucleic acids, including virus genomic DNA, in total DNA isolated from MRC-5 cells (ATCC CCL-171™?) infected with Human herpesvirus 5 (HCMV) strain Towne (ATCC VR-977?™).Virus and host cells are also available through the ATCC Catalog. This product was prepared using methods known to inactivate viruses.
Catalog No. NC2068808
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.