Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HGF Alexa Fluor Plus 555-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10113515
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10113515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10113515 Supplier R&D Systems Supplier No. FAB294AFP555100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

HGF Monoclonal antibody specifically detects HGF in Human samples. It is validated for Immunohistochemistry,Western Blot,Neutralization

Specifications

Antigen HGF
Applications Immunohistochemistry, Western Blot, Neutralization
Classification Monoclonal
Clone 24612
Conjugate Alexa Fluor Plus 555
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3082
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied
Product Type Antibody
Isotype IgG1
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.