Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human HGF Quantikine ELISA Kit
SDP

Catalog No. SHG00B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 Pack (6 Plates)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SHG00B 1 Pack (6 Plates)
DHG00B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SHG00B Supplier R&D Systems™ Supplier No. SHG00B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human HGF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human HGF Quantikine ELISA Kit quantifies human HGF in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), citrate plasma (50 ul).

1 Kit

Specifications

Assay Range 156.00 - 10000 pg/mL
Assay Sensitivity 24.1 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human HGF
For Use With (Application) ELISA
Gene ID (Entrez) 3082
Host Species Human
Quantity 1 Pack (6 Plates)
Sample Volume Cell Culture Supernates (50 uL),Serum (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL),Citrate Plasma (50 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A
Target HGF
Total Assay Time 4 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.