Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human HGF Recombinant Protein, PeproTech®

Catalog No. 10039H1MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
100-39H-1MG 1 mg
10039H100UG 100 μg
10039H250UG 250 μg
10039H25UG 25 μg
10039H500UG 500 μg
100-39H-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 100-39H-1MG Supplier Gibco™ Supplier No. 10039H1MG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human HGF, sourced from HEK293 cells, is a 79.4 kDa polypeptide consisting of 695 amino acid residues. As a result of glycosylation, Recombinant Human HGF migrates with an apparent molecular mass of approximately 68-85 kDa by SDS-PAGE gel, under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Hepatocyte growth factor regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signaling cascade after binding to the proto-oncogenic c-Met receptor. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. It is secreted as a single inactive polypeptide and is cleaved by serine proteases into a 69-kDa alpha-chain and 34-kDa beta-chain. A disulfide bond between the alpha and beta chains produces the active, heterodimeric molecule. The protein belongs to the plasminogen subfamily of S1 peptidases but has no detectable protease activity. Alternative splicing of this gene produces multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14210
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3082
Molecular Weight (g/mol) 79.4 kDa
Name Human HGF
Quantity 1 mg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF
Common Name HGF
Gene Symbol Hgf
Biological Activity Determined by the dose-dependent stimulation of the proliferation of monkey 4 MBr-5 cells. The expected ED50 for this effect is 1.0-20.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence Alpha chain: KRRNTIHEFK KSAKTTLIKI DPALKIKTKK VNTADQCANR CTRNKGLPFT CKAFVFDKAR KQCLWFPFNS MSSGVKKEFG HEFDLYENKD YIRNCIIGKG RSYKGTVSIT KSGIKCQPWS SMIPHEHSFL PSSYRGKDLQ ENYCRNPRGE EGGPWCFTSN PEVRYEVCDI PQCSEVECMT CNGESYRGLM DHTESGKICQ RWDHQTPHRH KFLPERYPDK GFDDNYCRNP DGQPRPWCYT LDPHTRWEYC AIKTCADNTM NDTDVPLETT ECIQGQGEGY RGTVNTIWNG IPCQRWDSQY PHEHDMTPEN FKCKDLRENY CRNPDGSESP WCFTTDPNIR VGYCSQIPNC DMSHGQDCYR GNGKNYMGNL SQTRSGLTCS MWDKNMEDLH RHIFWEPDAS KLNENYCRNP DDDAHGPWCY TGNPLIPWDY CPISRCEGDT TPTIVNLDHP VISCAKTKQL R Beta chain: VVNGIPTRTN IGWMVSLRYR NKHICGGSLI KESWVLTARQ CFPSRDLKDY EAWLGIHDVH GRGDEKCKQV LNVSQLVYGP EGSDLVLMKL ARPAVLDDFV STIDLPNYGC TIPEKTSCSV YGWGYTGLIN YDGLLRVAHL YIMGNEKCSQ HHRGKVTLNE SEICAGAEKI GSGPCEGDYG GPLVCEQHKM RMVLGVIVPG RGCAIPNRPG IFVRVAYYAK WIHKIILTYK VPQS
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.