Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HGFR/c-MET APC-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. FAB3582A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3582A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3582A Supplier R&D Systems Supplier No. FAB3582A

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

HGFR/c-MET Monoclonal specifically detects HGFR/c-MET in Human∣Xenograft samples. It is validated for Flow Cytometry∣Immunocytochemistry.

Specifications

Antigen HGFR/c-MET
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 95106
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P08581
Gene Alias AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met
Gene Symbols MET
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human HGF R/c-MET Glu25-Thr932 Accession # P08581
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4233
Test Specificity Detects human HGF R/c-MET.
Target Species Human, Xenograft
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.