Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HHATL Synthetic Peptide

Catalog No. PEP1398
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1398 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1398 Supplier Invitrogen™ Supplier No. PEP1398
Add to Cart
Add to Cart

Synthetic Peptide

HHATL Synthetic Peptide, PEP-1398, from Invitrogen.

HHATL (Hedgehog Acyltransferase-Like) is a gene that codes for a protein involved in the regulation of Hedgehog signaling pathways. HHATL is located on chromosome 7q36.3. Structurally, the protein encoded by this gene is characterized by an acyltransferase domain, which plays a crucial role in the N-palmitoylation of the Sonic Hedgehog (Shh) protein, a process essential for its proper signaling activity and distribution. Functionally, HHATL has been implicated in various biological processes, including amelioration of endoplasmic reticulum stress through autophagy. The gene is expressed in several tissues, with notable high expression in the heart, suggesting a potential role in cardiac development and function. Dysfunction in HHATL could lead to anomalies in Hedgehog signaling, impacting various developmental and cellular processes.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 57467
Species Human
Purification Method purified
Accession Number Q9HCP6
Common Name Human HHATL
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol HHATL
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.