Learn More
Kalen Biomedical LLC Human High Oxidized Low Density Lipoprotein, 1mg
Supplier:  Kalen Biomedical LLC 7702527
Encompass_Preferred
Purified human LDL (d=1.019 - 1.063 g/mL) is extensively oxidized with copper (II) sulfate to provide a High Oxidized LDL. It is dialyzed and ultrafiltered through a membrane and packaged aseptically under nitrogen. The product is provided in a solution with 154 mM NaCl, 5.6 mM Na2HPO4, 1.1 mM KH2PO4, and 0.34 mM EDTA, at pH 7.4.
Catalog No. NC0285115
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.