Learn More
R&D Systems™ Human High Sensitivity Cytokine Panel A Luminex™ Performance Control
Shop All Novus Products

Description
- Human High Sensitivity Cytokine Panel A Luminex™ Performance Control
- For use as quantitative controls for the determination of human cytokine concentrations in biological fluids
- Concentrations have been assigned using R&D Systems Luminex™ High Performance Assays
- Controls are prepared in a buffered protein solution with preservatives
- They contain recombinant human cytokines at low, medium and high concentrations
- Controls are supplied lyophilized
Specifications
Specifications
| Concentration | Control recombinant human cytokines at low, medium and high concentrations |
| Detects | Human cytokine concentrations in biological fluids |
| For Use With (Application) | For use with the Human High Senstivity Cytokine Panel A |
| Quantity | 1 |
| Source | Human |
| Content And Storage | Store the unopened product at 2 - 8°C. Do not use past expiration date. |
| Format | Controls are prepared in a buffered protein solution with preservatives. Controls are supplied lyophilized. Reconstitute each vial with the volume of Calibrator Diluent indicated on the product datasheet. |
| Product Line | R&D Systems |
- The ranges were determined using R&D Systems Luminex High Performance Assays. If expected values are not obtained, verify that the lot numbers on the vials correspond with the lot numbers listed on the product datasheet and the correct volume of deionized or distilled water was used for reconstitution of the controls.
- The results obtained with these controls depend upon several factors associated with methods and instrumentation. Test systems other than those supplied by R&D Systems may result in values that differ from those printed on this product datasheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.