Learn More
R&D Systems™ Human High Sensitivity Cytokine Panel B Luminex™ Performance Control
Shop All Novus Biologicals Products

Description
- For use as quantitative controls for the determination of human cytokine concentrations in biological fluids.
- Concentrations have been assigned using R&D Systems Luminex™ High Performance Assays.
- Controls are prepared in a buffered protein solution with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Ea. |
| Species | Human |
| Binding Target | Luminex Assay Controls |
| For Use With (Equipment) | Human Magnetic HS Cytokine B Panel |
| Product Line | R&D Systems |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.