Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-USP11 Protein, CF

Catalog No. E602050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E602050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E602050 Supplier R&D Systems™ Supplier No. E602050
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Reaction conditions will need to be optimized for each specific application. We recommend an initial recombinant human USP11 concentration of 10-50 nM when using Ubiquitin-AMC or Ubiquitin-Rhodamine substrates (U-550, U-555).

Specifications

Accession Number P51784
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 10% (v/v) Glycerol, 1 mM TCEP
Gene ID (Entrez) 8237
Molecular Weight (g/mol) M.W. (theoretical): 111 kDa
Name USP47
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP11 protein Contains an C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias Deubiquitinating enzyme 11, EC 3.1.2.15, EC 3.4.19.12, ubiquitin carboxyl-terminal hydrolase 11, ubiquitin specific peptidase 11, ubiquitin specific protease 11, ubiquitin thioesterase 11, Ubiquitin thiolesterase 11, ubiquitin-specific processing protease 11, Ubiquitin-specific-processing protease 11, UHX1, UHX1ubiquitin carboxyl-terminal hydrolase, X-linked
Common Name USP11
Gene Symbol USP11
Biological Activity Reaction conditions will need to be optimized for each specific application. We recommend an initial recombinant human USP11 concentration of 10-50 nM when using Ubiquitin-AMC or Ubiquitin-Rhodamine substrates (U-550, U-555).
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Contains an C-terminal 6-His tag
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein USP11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.