Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-USP12 Protein, CF

Catalog No. E578050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E578050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E578050 Supplier R&D Systems™ Supplier No. E578050
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Recombinant Human USP12 is a Ubiquitin-specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial USP12 concentration of 50-500 nM. Requires a 1:1 stoichiometric amount of UAF1 (E-566) for activity when using Ubiquitin-AMC or Ubiquitin-Rhodamine (U-550, U-555) as a substrate. In vitro, the USP12/UAF1 complex will not efficiently hydrolyze poly-Ubiquitin chains in the absence of WDR20.

Specifications

Accession Number O75317
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 8.2, 200 mM NaCl, 10% (v/v) Glycerol, 2 mM TCEP
Gene ID (Entrez) 219333
Molecular Weight (g/mol) M.W. (theoretical): 44 kDa
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP12 protein Contains a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias Deubiquitinating enzyme 12, EC 3.1.2.15, EC 3.4.19.12, UBH1, UBH1ubiquitin specific protease 12, ubiquitin carboxyl-terminal hydrolase 12, ubiquitin specific peptidase 12, ubiquitin specific protease 12 like 1, ubiquitin thioesterase 12, Ubiquitin thiolesterase 12, Ubiquitin-hydrolyzing enzyme 1, Ubiquitin-specific-processing protease 12, USP12L1
Common Name USP12
Gene Symbol USP12
Biological Activity Recombinant Human USP12 is a Ubiquitin-specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial USP12 concentration of 50-500 nM. Requires a 1:1 stoichiometric amount of UAF1 (E-566) for activity when using Ubiquitin-AMC or Ubiquitin-Rhodamine (U-550, U-555) as a substrate. In vitro, the USP12/UAF1 complex will not efficiently hydrolyze poly-Ubiquitin chains in the absence of WDR20.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Contains a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein USP12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.