Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-USP13 Protein, CF

Catalog No. E588050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E588050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E588050 Supplier R&D Systems™ Supplier No. E588050
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human USP13 concentration of 100-500 nM when using Ubiquitin-AMC or Ubiquitin-Rh110 substrates (U-550, U-555)

Specifications

Accession Number Q92995
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 10% (v/v) Glycerol, 1 mM TCEP
Gene ID (Entrez) 8975
Molecular Weight (g/mol) M.W. (theoretical): 98 kDa
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP13 protein Contains an C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias Deubiquitinating enzyme 13, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T-3, IsoT-3, ISOT3IsoT-3, ubiquitin carboxyl-terminal hydrolase 13, ubiquitin specific peptidase 13 (isopeptidase T-3), ubiquitin specific protease 13 (isopeptidase T-3), ubiquitin thioesterase 13, Ubiquitin thiolesterase 13, Ubiquitin-specific-processing protease 13
Common Name USP13
Gene Symbol USP13
Biological Activity Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human USP13 concentration of 100-500 nM when using Ubiquitin-AMC or Ubiquitin-Rh110 substrates (U-550, U-555)
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Contains an C-terminal 6-His tag
Purity or Quality Grade >98%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Protein USP13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.