Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His6-VCIP135/VCPIP1 Protein, CF

Catalog No. E590050
Change view
Click to view available options
50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
E590050 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E590050 Supplier R&D Systems™ Supplier No. E590050
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Reaction conditions will need to be optimized for each specific application. We recommend an initial VCIP135 concentration of 100-500 nM in reactions utilizing Ubiquitin-AMC or Ubiquitin-Rhodamine substrates (U-550 or U-555) or Poly-ubiquitin chain substrates.

Specifications

Accession Number Q96JH7
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 8.0, 100 mM NaCl, 10% (v/v) Glycerol, 1 mM TCEP
Gene ID (Entrez) 80124
Molecular Weight (g/mol) M.W. (theoretical): 135 kDa
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human VCIP135/VCPIP1 protein Contains an C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias DKFZp686G038, DUBA3, EC 3.4.22.-, FLJ23132, KIAA1850FLJ60694, valosin containing protein (p97)/p47 complex interacting protein 1, valosin-containing protein (p97)/p47 complex-interacting protein p135, Valosin-containing protein p97/p47 complex-interacting protein 1, Valosin-containing protein p97/p47 complex-interacting protein p135, VCIP135deubiquitinating protein VCIP135, VCPIP1, VCPIP1 VCIP135
Common Name VCIP135/VCPIP1
Gene Symbol VCPIP1
Biological Activity Reaction conditions will need to be optimized for each specific application. We recommend an initial VCIP135 concentration of 100-500 nM in reactions utilizing Ubiquitin-AMC or Ubiquitin-Rhodamine substrates (U-550 or U-555) or Poly-ubiquitin chain substrates.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Contains an C-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein VCIP135/VCPIP1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.