Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys-Ser-Thr-Gly-Gly-Lys-Ala-Pro-Arg-Lys-Gln-Leu-Cys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HIST001A
Encompass
ONE-LETTER SEQUENCE: ARTKQTARKSTGGKAPRKQLCMOLECULAR FORMULA: C94H172N36O28S1MOLECULAR WEIGHT:2286.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9422
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.