Learn More
Abcam Human Histone H3 (di Methyl K36) Peptide, >90% Purity, 1 mg
Description
- Target Protein: Histone H3
- Purity: >90%
- Tested Applications: BL
- Form: Liquid
- Storage Buffer: HEPES, 0.87% Sodium chloride, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Reactivity | Human |
| Shipping Condition | Blue Ice |
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Histone H3 (di methyl K36), Human |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Buffer | HEPES, 0.87% sodium chloride, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated |
| Cross Reactivity | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.