Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human Histone H3 Peptide, Synthetic, >70% Purity, 200 μL

Catalog No. 502708553
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-8553 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-8553 Supplier Abcam Supplier No. AB94338200UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic human Histone H3 peptide with >70% purity, suitable for functional studies and inhibition assays.

  • Protein Name: Histone H3
  • Species Reactivity: Human
  • Form: Liquid
  • Purity: >70%
  • Applications: FuncS, Inhib
  • Storage Conditions: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Form Liquid
Reactivity Human
Substrate Properties Peptide-based Substrate
Target Molecule Histone H3 (Human)
Buffer pH7
Label or Dye Unconjugated
Shipping Condition Dry Ice
Formulation pH 7
Quantity 200 μL
For Use With (Application) FuncS, Inhib
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.