Learn More
Abcam Human Histone H3 Peptide, Unmodified, Lyophilized, 500 μg
Description
Human Histone H3 (unmodified) peptide is a synthetic blocking peptide.
- Species Reactivity: Human
- Tested Applications: Dot, BL
- Purity: >90%
- Form: Lyophilized
- Citation Count: 6
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Target Molecule | Histone H3 (Human) |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Substrate Properties | Peptide-based Substrate |
| Label or Dye | Unconjugated |
| Cross Reactivity | Human |
| Quantity | 500 μg |
| For Use With (Application) | Blocking, Dot Blot, Western Blot |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.