Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Human Histone H3 Tri Methyl K9 Peptide, Lyophilized, >90% Purity, 1 mg

Catalog No. 502704431
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4431 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4431 Supplier Abcam Supplier No. AB17731MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide with >90% purity, suitable for Western blot applications.

  • Applications: BL
  • Purity: >90%
  • Form: Lyophilized
  • Species: Human
  • Product Type: Synthetic blocking peptide

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Peptide-based Substrate
Target Molecule Histone H3 (tri methyl K9)
Label or Dye Unconjugated
Reactivity Human
Shipping Condition Blue Ice
Content And Storage Store at -20°C.
Form Lyophilized
Detection Location In-Blot Detection
Cross Reactivity Human
Quantity 1 mg
For Use With (Application) Blocking, Western Blot
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.