Learn More
CPC Scientific H-Ala-Gly-Gly-Lys-Gly-Gly-Lys-Gly-Met-Gly-Lys-Val-Gly-Ala-Lys-Arg-His-Ser-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST002B
Encompass
ONE-LETTER SEQUENCE: AGGKGGKGMGKVGAKRHSMOLECULAR FORMULA: C69H123N27O20S1MOLECULAR WEIGHT:1683STORAGE CONDITIONS: -20 5CRESEARCH AREA: Miscellaneous
Catalog No. 50-210-9425
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.