Learn More
ACROBiosystems Human HLA-A0201B2Mp53 HMT
Supplier:  ACROBiosystems HLPH52H5200UG
Encompass_Preferred
Human HLA-A*0201B2Mp53 (HMTEVVRRC) Tetramer Protein (HLP-H52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) HMTEVVRRC peptide (Accession # AAA59606.1 (HLA-A*0201) P61769-1 (B2M) HMTEVVRRC).
Catalog No. 50-253-4701
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.