Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
| Assay Range | 62.5 to 4000 pg/mL |
| Assay Sensitivity | 37.5 pg/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Human HLA B in samples. No significant cross-reactivity or interference between Human HLA B and analogues was observed. |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3106 |
| Gene Symbol | HLA-B |
| Quantity | 1 Kit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
