Learn More
Sino Biological Human HMGB1 / HMG1 Protein (His Tag) 500ug
Supplier:  Sino Biological 10326H08H500
Encompass_Preferred
A DNA sequence encoding the human HMGB1 protein (NP_002119.1) (Met 1-Glu 215) was fused with a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus.
Catalog No. 50-255-5382
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.