Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HMGN3 Full-length ORF (AAH09529, 1 a.a. - 77 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947644 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-644 25 μg
89-947-643 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-947-644 Supplier Abnova™ Supplier No. H00009324P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Thyroid hormone receptors are hormone-dependent transcription factors that regulate expression of a variety of specific target genes. The protein encoded by this gene binds thyroid hormone receptor beta, but only in the presence of thyroid hormone. The encoded protein, a member of the HMGN protein family, is thought to reduce the compactness of the chromatin fiber in nucleosomes, thereby enhancing transcription from chromatin templates. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MPKRKSPENTEGKDGSKVTKQEPTRRSARLSAKPAPPKPEPKPRKTSAKKEPGAKISRGAKGKKEEKQEAGKEGTEN

Specifications

Accession Number AAH09529
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9324
Molecular Weight (g/mol) 34.21kDa
Name HMGN3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MPKRKSPENTEGKDGSKVTKQEPTRRSARLSAKPAPPKPEPKPRKTSAKKEPGAKISRGAKGKKEEKQEAGKEGTEN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686E20226/PNAS-24/PNAS-25/TRIP7
Common Name HMGN3
Gene Symbol HMGN3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.