Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human HMW Adiponectin/Acrp30 Quantikine ELISA Kit

Catalog No. DHWAD0
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
DHWAD0 1 Plate 1 x 96-well Plate
SHWAD0 6 Plates 6 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DHWAD0 Supplier R&D Systems™ Supplier No. DHWAD0
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human HMW Adiponectin/Acrp30 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human HMW Adiponectin/Acrp30 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Adiponectin/Acrp30 in serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 3.90 - 250ng/mL
Assay Sensitivity 0.989ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human high molecular weight Adiponectin
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 9370
Quantity 1 Plate
Sample Volume Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ACDC, Acrp30, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, ApM1, APM1APM-1, C1q and collagen domain-containing protein, GBP28, GBP28apM1, Gelatin-binding protein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.