Learn More
Alpco Ltd ALPCO Human HMW & Total Adiponectin ELISA, Research Use Only, Sensitive Measurement Range 0.078-5 ng/mL, 96 Wells
SDP
Supplier:  Alpco Ltd 80ADPHUE01
Encompass_Preferred
The ALPCO HMW & Total Adiponectin ELISA is a highly specific, quantitative enzyme-linked immunosorbent assay for measuring both high molecular weight and total adiponectin in human serum and plasma. It provides accurate differentiation with high sensitivity (<0.1 ng/mL), making it essential for research in metabolic conditions like Type 2 Diabetes and obesity. Its sandwich assay design and no cross-reactivity with similar proteins ensure reliable results for researchers.
Catalog No. NC1836978
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.