Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOOK1 Full-length ORF (NP_056972.1, 1 a.a. - 728 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947713 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-713 10 μg
89-947-714 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-947-713 Supplier Abnova™ Supplier No. H00051361P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the hook family of coiled-coil proteins, which bind to microtubules and organelles through their N- and C-terminal domains, respectively. The encoded protein localizes to discrete punctuate subcellular structures, and interacts with several members of the Rab GTPase family involved in endocytosis. It is thought to link endocytic membrane trafficking to the microtubule cytoskeleton. Several alternatively spliced transcript variants have been identified, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: MEETQPPPQPKLPLCDSLMIWLQTFNTASPCQDVKQLTSGVAMAQVLHQIDAAWFNESWLSRIKEDVGDNWRIKASNVKKVLQGIMSYYHEFLGQQISEALIPDLNQITECSDPVELGRLLQLILGCAINCEKKQEHIQNIMTLEESVQHVVMTAIQELMSKEILSSPPNDAVGELEQQLKRALEELQEALAEKEELRQRCEELDMQVTTLQDEKNSLVSENEMMNEKLDQLDGSFDDPNTVVAKKYFHAQLQLEQLQEENFRLEAAKDDYRVHCEELEKQLIEFQHRNDELTSLAEETRALKDEIDVLRATSDKANKLESTVEIYRQKLQDLNDLRKQVKTLQETNMMYMHNTVSLEEELKKANAARTQLETYKRQVQDLHVKLSSESKRADTLAFEMKRLEEKHEALLKEKERLIEQRDTLKETNEELRCSQVQQDHLNQTDASATKSYENLAAEIMPVEYREVFIRLQHENKMLRLQQEGSENERIEELQEQLEQKHRKMNELETEQRLSKERIRELQQQIEDLQKSLQEQGSKSEGESSSKLKQKLEAHMEKLTEVHEELQKKQELIEDLQPDINQNVQKINELEAALQKKDEDMKAMEERYKMYLEKARNVIKTLDPKLNPASAEIMLLRKQLAEKERRIEILESECKVAKFRDYEEKLIVSAWYNKSLAFQKLGMESRLVSGGGACSDTGACTPARSFLAQQRHITNTRRNLSVKVPATTSD

Specifications

Accession Number NP_056972.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51361
Molecular Weight (g/mol) 111kDa
Name HOOK1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MEETQPPPQPKLPLCDSLMIWLQTFNTASPCQDVKQLTSGVAMAQVLHQIDAAWFNESWLSRIKEDVGDNWRIKASNVKKVLQGIMSYYHEFLGQQISEALIPDLNQITECSDPVELGRLLQLILGCAINCEKKQEHIQNIMTLEESVQHVVMTAIQELMSKEILSSPPNDAVGELEQQLKRALEELQEALAEKEELRQRCEELDMQVTTLQDEKNSLVSENEMMNEKLDQLDGSFDDPNTVVAKKYFHAQLQLEQLQEENFRLEAAKDDYRVHCEELEKQLIEFQHRNDELTSLAEETRALKDEIDVLRATSDKANKLESTVEIYRQKLQDLNDLRKQVKTLQETNMMYMHNTVSLEEELKKANAARTQLETYKRQVQDLHVKLSSESKRADTLAFEMKRLEEKHEALLKEKERLIEQRDTLKETNEELRCSQVQQDHLNQTDASATKSYENLAAEIMPVEYREVFIRLQHENKMLRLQQEGSENERIEELQEQLEQKHRKMNELETEQRLSKERIRELQQQIEDLQKSLQEQGSKSEGESSSKLKQKLEAHMEKLTEVHEELQKKQELIEDLQPDINQNVQKINELEAALQKKDEDMKAMEERYKMYLEKARNVIKTLDPKLNPASAEIMLLRKQLAEKERRIEILESECKVAKFRDYEEKLIVSAWYNKSLAFQKLGMESRLVSGGGACSDTGACTPARSFLAQQRHITNTRRNLSVKVPATTSD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HK1/MGC10642
Common Name HOOK1
Gene Symbol HOOK1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.