Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOXA10 Full-length ORF (ABM86018.1, 1 a.a. - 393 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947732 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-732 25 ug
89-947-731 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-947-732 Supplier Abnova™ Supplier No. H00003206P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor that may regulate gene expression, morphogenesis, and differentiation. More specifically, it may function in fertility, embryo viability, and regulation of hematopoietic lineage commitment. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]

Sequence: MSCSESPAANSFLVDSLISSGRGEAGGGGGGAGGGGGGGYYAHGGVYLPPAADLPYGLQSCGLFPTLGGKRNEAASPGSGGGGGGLGPGAHGYGPSPIDLWLDAPRSCRMEPPDGPPPPPQQQPPPPPQPPQPAPQATSCSFAQNIKEESSYCLYDSADKCPKVSATAAELAPFPRGPPPDGCALGTSSGVPVPGYFRLSQAYGTAKGYGSGGGGAQQLGAGPFPAQPPGRGFDLPPALASGSADAARKERALDSPPPPTLACGSGGGSQGDEEAHASSSAAEELSPAPSESSKASPEKDSLGNSKGENAANWLTAKSGRKKRCPYTKHQTLELEKEFLFNMYLTRERRLEISRSVHLTDRQVKIWFQNRRMKLKKMNRENRIRELTANFNFS

Specifications

Accession Number ABM86018.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3206
Molecular Weight (g/mol) 69.63kDa
Name HOXA10 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 25 ug
Immunogen MSCSESPAANSFLVDSLISSGRGEAGGGGGGAGGGGGGGYYAHGGVYLPPAADLPYGLQSCGLFPTLGGKRNEAASPGSGGGGGGLGPGAHGYGPSPIDLWLDAPRSCRMEPPDGPPPPPQQQPPPPPQPPQPAPQATSCSFAQNIKEESSYCLYDSADKCPKVSATAAELAPFPRGPPPDGCALGTSSGVPVPGYFRLSQAYGTAKGYGSGGGGAQQLGAGPFPAQPPGRGFDLPPALASGSADAARKERALDSPPPPTLACGSGGGSQGDEEAHASSSAAEELSPAPSESSKASPEKDSLGNSKGENAANWLTAKSGRKKRCPYTKHQTLELEKEFLFNMYLTRERRLEISRSVHLTDRQVKIWFQNRRMKLKKMNRENRIRELTANFNFS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HOX1/HOX1.8/HOX1H/MGC12859/PL
Common Name HOXA10
Gene Symbol HOXA10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.