Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOXA7 Full-length ORF (NP_008827.2, 1 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947751 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-751 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-947-751 Supplier Abnova™ Supplier No. H00003204P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. For example, the encoded protein represses the transcription of differentiation-specific genes during keratinocyte proliferation, but this repression is then overcome by differentiation signals. This gene is highly similar to the antennapedia (Antp) gene of Drosophila. [provided by RefSeq]

Sequence: MSSSYYVNALFSKYTAGASLFQNAEPTSCSFAPNSQRSGYGAGAGAFASTVPGLYNVNSPLYQSPFASGYGLGADAYGNLPCASYDQNIPGLCSDLAKGACDKTDEGALHGAAEANFRIYPWMRSSGPDRKRGRQTYTRYQTLELEKEFHFNRYLTRRRRIEIAHALCLTERQIKIWFQNRRMKWKKEHKDEGPTAAAAPEGAVPSAAATAAADKADEEDDDEEEEDEEE

Specifications

Accession Number NP_008827.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3204
Molecular Weight (g/mol) 51.8kDa
Name HOXA7 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MSSSYYVNALFSKYTAGASLFQNAEPTSCSFAPNSQRSGYGAGAGAFASTVPGLYNVNSPLYQSPFASGYGLGADAYGNLPCASYDQNIPGLCSDLAKGACDKTDEGALHGAAEANFRIYPWMRSSGPDRKRGRQTYTRYQTLELEKEFHFNRYLTRRRRIEIAHALCLTERQIKIWFQNRRMKWKKEHKDEGPTAAAAPEGAVPSAAATAAADKADEEDDDEEEEDEEE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ANTP/HOX1/HOX1.1/HOX1A
Common Name HOXA7
Gene Symbol HOXA7
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.