Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOXB13 Full-length ORF (AAH07092, 1 a.a. - 284 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947760 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-760 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-947-760 Supplier Abnova™ Supplier No. H00010481P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a transcription factor that belongs to the homeobox gene family. Genes of this family are highly conserved among vertebrates and essential for vertebrate embryonic development. This gene has been implicated to play a role in fetal skin development and cutaneous regeneration. In mice, a similar gene was shown to exhibit temporal and spatial colinearity in the main body axis of the embryo, but was not expressed in the secondary axes, which suggests functions in body patterning along the axis. This gene and other HOXB genes form a gene cluster at chromosome the 17q21-22 region. [provided by RefSeq]

Sequence: MEPGNYATLDGAKDIEGLLGAGGGRNLVAHSPLTSHPAAPTLMPAVNYAPLDLPGSAEPPKQCHPCPGVPQGTSPAPVPYGYFGGGYYSCRVSRSSLKPCAQAATLAAYPAETPTAGEEYPSRPTEFAFYPGYPGTYQPMASYLDVSVVQTLGAPGEPRHDSLLPVDSYQSWALAGGWNSQMCCQGEQNPPGPFWKAAFADSSGQHPPDACAFRRGRKKRIPYSKGQLRELEREYAANKFITKDKRRKISAATSLSERQITIWFQNRRVKEKKVLAKVKNSATP

Specifications

Accession Number AAH07092
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10481
Molecular Weight (g/mol) 56.98kDa
Name HOXB13 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MEPGNYATLDGAKDIEGLLGAGGGRNLVAHSPLTSHPAAPTLMPAVNYAPLDLPGSAEPPKQCHPCPGVPQGTSPAPVPYGYFGGGYYSCRVSRSSLKPCAQAATLAAYPAETPTAGEEYPSRPTEFAFYPGYPGTYQPMASYLDVSVVQTLGAPGEPRHDSLLPVDSYQSWALAGGWNSQMCCQGEQNPPGPFWKAAFADSSGQHPPDACAFRRGRKKRIPYSKGQLRELEREYAANKFITKDKRRKISAATSLSERQITIWFQNRRVKEKKVLAKVKNSATP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PSGD
Common Name HOXB13
Gene Symbol HOXB13
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.