Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOXC13 Partial ORF (NP_059106, 132 a.a. - 231 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947793 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-793 25 ug
89-947-792 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-947-793 Supplier Abnova™ Supplier No. H00003229Q01L

Used for AP, Array, ELISA, WB-Re

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the development of hair, nail, and filiform papilla. [provided by RefSeq]

Sequence: CRLSHNVNLQQKPCAYHPGDKYPEPSGALPGDDLSSRAKEFAFYPSFASSYQAMPGYLDVSVVPGISGHPEPRHDALIPVEGYQHWALSNGWDSQVYCSK

Specifications

Accession Number NP_059106
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3229
Molecular Weight (g/mol) 36.74kDa
Name HOXC13 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen CRLSHNVNLQQKPCAYHPGDKYPEPSGALPGDDLSSRAKEFAFYPSFASSYQAMPGYLDVSVVPGISGHPEPRHDALIPVEGYQHWALSNGWDSQVYCSK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HOX3/HOX3G
Common Name HOXC13
Gene Symbol HOXC13
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.