Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOXD1 Full-length ORF (AAH14477.1, 1 a.a. - 328 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947809 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-809 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-947-809 Supplier Abnova™ Supplier No. H00003231P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the Antp homeobox family and encodes a protein with a homeobox DNA-binding domain. This nuclear protein functions as a sequence-specific transcription factor that is involved in differentiation and limb development. Mutations in this gene have been associated with severe developmental defects on the anterior-posterior (a-p) limb axis. [provided by RefSeq]

Sequence: MSSYLEYVSCSSSGGVGGDVLSLAPKFCRSDARPVALQPAFPLGNGDGAFVSCLPLAAARPSPSPPAAPARPSVPPPAAPQYAQCTLEGAYEPGAAPAAAAGGADYGFLGSGPAYDFPGVLGRAADDGGSHVHYATSAVFSGGGSFLLSGQVDYAAFGEPGPFSACLKASADGHPGAFQTASPAPGTYPKSVSPASGLPAAFSTFEWMKVKRNASKKGKLAEYGAASPSSAIRTNFSTKQLTELEKEFHFNKYLTRARRIEIANCLHLNDTQVKIWFQNRRMKQKKREREGLLATAIPVAPLQLPLSGTTPTKFIKNPGSPSQSQEPS

Specifications

Accession Number AAH14477.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3231
Molecular Weight (g/mol) 60.5kDa
Name HOXD1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MSSYLEYVSCSSSGGVGGDVLSLAPKFCRSDARPVALQPAFPLGNGDGAFVSCLPLAAARPSPSPPAAPARPSVPPPAAPQYAQCTLEGAYEPGAAPAAAAGGADYGFLGSGPAYDFPGVLGRAADDGGSHVHYATSAVFSGGGSFLLSGQVDYAAFGEPGPFSACLKASADGHPGAFQTASPAPGTYPKSVSPASGLPAAFSTFEWMKVKRNASKKGKLAEYGAASPSSAIRTNFSTKQLTELEKEFHFNKYLTRARRIEIANCLHLNDTQVKIWFQNRRMKQKKREREGLLATAIPVAPLQLPLSGTTPTKFIKNPGSPSQSQEPS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HOX4/HOX4G/Hox-4.7
Common Name HOXD1
Gene Symbol HOXD1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.