Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human HPRG Recombinant Protein, PeproTech®

Catalog No. 10060250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10060250UG 250 μg
10060100UG 100 μg
100-60-1MG 1 mg
100-60-25UG 25 μg
10060500UG 500 μg
100-60-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 10060250UG Supplier Gibco™ Supplier No. 10060250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human HPRG is a 57.7 kDa glycoprotein containing 507 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein's effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04196
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3273
Molecular Weight (g/mol) 57.7 kDa
Name Human HPRG
Quantity 250 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AI265597; AW413091; D16jh2; D18020; DKFZp779H1622; histidine rich glycoprotein; histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; histidine-rich glycoprotein 1; HPRG; HRG; HRG1; Hrgp; THPH11
Common Name HRG
Gene Symbol HRG
Biological Activity Measured by its ability to support adhesion of Con-A activated MOLT-4 cells to the Recombinant Human HPRG-coated surface. The ED50 for this effect is 1.5-2.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence VSPTDCSAVE PEAEKALDLI NKRRRDGYLF QLLRIADAHL DRVENTTVYY LVLDVQESDC SVLSRKYWND CEPPDSRRPS EIVIGQCKVI ATRHSHESQD LRVIDFNCTT SSVSSALANT KDSPVLIDFF EDTERYRKQA NKALEKYKEE NDDFASFRVD RIERVARVRG GEGTGYFVDF SVRNCPRHHF PRHPNVFGFC RADLFYDVEA LDLESPKNLV INCEVFDPQE HENINGVPPH LGHPFHWGGH ERSSTTKPPF KPHGSRDHHH PHKPHEHGPP PPPDERDHSH GPPLPQGPPP LLPMSCSSCQ HATFGTNGAQ RHSHNNNSSD LHPHKHHSHE QHPHGHHPHA HHPHEHDTHR QHPHGHHPHG HHPHGHHPHG HHPHGHHPHC HDFQDYGPCD PPPHNQGHCC HGHGPPPGHL RRRGPGKGPR PFHCRQIGSV YRLPPLRKGE VLPLPEANFP SFPLPHHKHP LKPDNQPFPQ SVSESCPGKF KSGFPQVSMF FTHTFPK
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.