Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HS3ST1 Partial ORF (NP_005105.1, 111 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89947907 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-947-907 10 μg
89-947-908 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-947-907 Supplier Abnova™ Supplier No. H00009957Q01S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for AP, Array, ELISA, WB-Re

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It possesses both heparan sulfate glucosaminyl 3-O-sulfotransferase activity, anticoagulant heparan sulfate conversion activity, and is a rate limiting enzyme for synthesis of anticoagulant heparan. This enzyme is an intraluminal Golgi resident protein. [provided by RefSeq]

Sequence: WPHQLTVEKTPAYFTSPKVPERVYSMNPSIRLLLILRDPSERVLSDYTQVFYNHMQKHKPYPSIEEFLVRDGRLNVDYKALNRSLYHVHMQNWLRFFPLR

Specifications

Accession Number NP_005105.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9957
Molecular Weight (g/mol) 36.74kDa
Name HS3ST1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen WPHQLTVEKTPAYFTSPKVPERVYSMNPSIRLLLILRDPSERVLSDYTQVFYNHMQKHKPYPSIEEFLVRDGRLNVDYKALNRSLYHVHMQNWLRFFPLR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias 3OST/3OST1
Common Name HS3ST1
Gene Symbol HS3ST1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.