Learn More
Human HSP47 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Hsp47 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 950828 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 5-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P50454 |
| Gene Alias | (collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, BERF-1, CBP1, CBP2, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), OI10, PPROM, proliferation-inducing gene 14, RA-A47, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH1, SERPINH2colligin |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.