Learn More
Specifications
Specifications
| Assay Range | 0.63 - 40 ng/mL |
| Product Type | Solid Phase Sandwich ELISA |
| Gene ID (Entrez) | 3326 |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | D6S182, Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90AB1, HSP90B, HSP90-BETA, HSP90BHSP 90, HSPC2, HSPC2D6S182, HSPCB, HSPCBFLJ26984 |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
