Learn More
Sino Biological Human HSPA5 PAb 100UL 102056-T40-100
Supplier:  Sino Biological 102056T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GRP78 / HSPA5, and purified by antigen affinity chromatography.
Catalog No. 50-240-9274
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.