Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HSPG2 Partial ORF (NP_005520.3, 25 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89948090 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-090 10 ug
89-948-091 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-948-090 Supplier Abnova™ Supplier No. H00003339Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Heparan sulfate proteoglycan is a major component of basement membranes, where the molecule may be involved in the stabilization of other molecules as well as being involved with glomerular permeability to macromolecules and cell adhesion. This form of HSPG, known as HSPG2 or perlecan, is encoded by a gene that maps to chromosome 1. The gene for the form of HSPG associated with the cell surface of fibroblasts has been mapped to human chromosome 8 (MIM 142460).[supplied by OMIM]

Sequence: GLRAYDGLSLPEDIETVTASQMRWTHSYLSDDEDMLADSISGDDLGSGDLGSGDFQMVYFRALVNFTRSIEYSPQLEDAGSREFREVSEAVVDTLESEYLKIPGDQVVSV

Specifications

Accession Number NP_005520.3
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3339
Molecular Weight (g/mol) 37.84kDa
Name HSPG2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen GLRAYDGLSLPEDIETVTASQMRWTHSYLSDDEDMLADSISGDDLGSGDLGSGDFQMVYFRALVNFTRSIEYSPQLEDAGSREFREVSEAVVDTLESEYLKIPGDQVVSV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PLC/PRCAN/SJA/SJS/SJS1
Common Name HSPG2
Gene Symbol HSPG2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.