Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human HTRA2/Omi (NP_037379.1) VersaClone cDNA
SDP

Catalog No. RDC3036 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC3036 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC3036 Supplier R&D Systems™ Supplier No. RDC3036
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

Specifications

Gene HTRA2/Omi
Accession Number NP_037379.1
Gene ID (Entrez) 27429
Gene Alias High temperature requirement protein A2, HtrA serine peptidase 2, HtrA2, HtrA-like serine protease, Omi, OMIOmi stress-regulated endoprotease, PARK13EC 3.4.21.108, PRSS25, PRSS25serine, 25, Serine protease 25, serine protease HTRA2, mitochondrial, Serine proteinase OMI
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.