Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human HVEM Fc Recombinant Protein, PeproTech®

Catalog No. 31027500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 100 μg
20 μg
5 x 100 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-027-500 5 x 100 μg
31027100UG 100 μg
31-027-200 2 x 100 μg
310-27-20UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31-027-500 Supplier Gibco™ Supplier No. 31027500
Add to Cart
Add to Cart

Recombinant Protein

310-27-1MG will be provided as 10 x 100 μg (310-27-100UG). 310-27-200UG will be provided as 2 x 100 μg (310-27-100UG). 310-27-500UG will be provided as 5 x 100 μg (310-27-100UG). Recombinant Human HVEM-Fc Chimera is a 376 amino acid fusion protein that contains an N-terminal domain corresponding to the extracellular region of HVEM, and a C-terminal domain corresponding to residues 102 to 330 of human IgG1. The calculated molecular weight of Recombinant Human HVEM-Fc is 41.4 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TNFRSF14 is a member of the TNF-receptor superfamily. TNFRSF14 was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to TNFRSF14 has been shown to be part of the viral entry mechanism. The cytoplasmic region of TNFRSF14 was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. Activation of the signal transduction pathway involving TNFRSF14 in T cells stimulates T cell proliferation and cytokine production, leading to inflammation and enhanced CTL-mediated tumor immunity, suggesting that these proteins may be useful as potential targets for controlling cellular immune responses. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these TNFRSF14 variants have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92956
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 8764
Molecular Weight (g/mol) 41.4 kDa
Name Human HVEM Fc
Purification Method Purified
Quantity 5 x 100 μg
Source Insect cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Atar; CD270; CD40-like protein; herpes virus entry mediator; herpes virus entry mediator A; Herpesvirus entry mediator A; HGNC:11912; HVEA; Hvem; LIGHTR; RP3-395M20.6; sCD2710; TNF receptor superfamily member 14; Tnfrs14; TNFRSF14; TR2; tumo; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2; UNQ329/PRO509
Common Name TNFRSF14 (HVEM)
Gene Symbol TNFRSF14
Biological Activity Determined by its ability to neutralize 0.25 ng/ml of hTNFbeta induced cytotoxicity on murine L929 cells. The expected ED50 for this effect is 1.3-1.9 ug/ml of HVEM-Fc.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence LPSCKEDEYP VGSECCPKCS PGYRVKEACG ELTGTVCEPC PPGTYIAHLN GLSKCLQCQM CDPAMGLRAS RNCSRTENAV CGCSPGHFCI VQDGDHCAAC RAYATSSPGQ RVQKGGTESQ DTLCQNCPPG TFSPNGTLEE CQHQTKRSCD KTHTCPPCPA PELLGGPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDP EVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPAP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGK
Content And Storage -20°C
Activity Determined by its ability to neutralize 0.25 ng/ml of hTNFbeta induced cytotoxicity on murine L929 cells. The expected ED50 for this effect is 1.3-1.9 ug/ml of HVEM-Fc.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.