Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human I-TAC (CXCL11), Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF300461MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF300461MG 1 mg
AF30046100 100 μg
AF3004620UG 20 μg
AF30046500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF300461MG Supplier Gibco™ Supplier No. AF300461MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human I-TAC (CXCL11) is an 8.3 kDa protein containing 73 amino acid residues, including the four highly conserved cysteine residues present in CXC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interferon-inducible T-cell alpha chemoattractant ( I-TAC), belonging to the CXC chemokine family, is expressed in peripheral blood leukocytes, pancreas and liver, thymus, spleen, lung, small intestine, placenta and prostate. Its molecular weight is 8-11 kD. I-TAC is also known as CXCL-11 and IP-9. The cytokine is involved in regulating calcium release and inducing a chemotactic response in activated T-cells and is the dominant ligand for CXC receptor-3. The gene encoding this protein contains 4 exons and at least three polyadenylation signals which might reflect cell-specific regulation of expression. IFN-gamma is a potent inducer of transcription of this gene. It is upregulated in patients with AIDS, dementia and in patients with multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14625
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 6373
Molecular Weight (g/mol) 8.3 kDa
Name Human I-TAC (CXCL11), Animal-Free
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias betaR1; beta-R1; b-R1; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 11; CXC; CXC motif chemokine; C-X-C motif chemokine 11; C-X-C motif chemokine ligand 11; Cxc11; CXCL; CXCL11; H174; Interferon gamma-inducible protein 9; interferon-inducible T-cell alpha chemoattractant; IP9; IP-9; Itac; I-TAC; MGC102770; Scyb11; Scyb9b; small inducible cytokine B11; small inducible cytokine subfamily B (Cys-X-Cys), member 11; small inducible cytokine subfamily B (Cys-X-Cys), member 9 B; small-inducible cytokine B11
Common Name CXCL11
Gene Symbol CXCL11
Biological Activity Determined by its ability to chemoattract human T-cells activated with IL-2.
Conjugate Unconjugated
Recombinant Recombinant
Sequence FPMFKRGRCL CIGPGVKAVK VADIEKASIM YPSNNCDKIE VIITLKENKG QRCLNPKSKQ ARLIIKKVER KNF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.