Learn More
R&D Systems™ Human ICAM-1/CD54 Allele-specific Quantikine ELISA Kit
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Assay Range | 1.56 - 50ng/mL |
| Assay Sensitivity | 0.254ng/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Line | R&D Systems |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Human sICAM-1 |
| Sufficient For | 6 x 96-well Plates |
| Target Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.